Merck
Binding modes and interaction mechanism between different base pairs and methylene blue trihydrate: a quantum mechanics study.
Advances in experimental medicine and biology (2014-11-13)
Peijun Xu, Jinguang Wang, Yong Xu, Huiying Chu, Hujun Shen, Depeng Zhang, Meixia Zhao, Jiahui Liu, Guohui Li
ABSTRAKT
Different quantum mechanic methods have been evaluated for the calculation of binding modes and interactions between intercalators with different DNA base pairs by comparing with the results of MP2, which is very expensive, indicating that WB97XD method under 6-311+G* basis set is able to efficiently reproduce MP2 results. We discovered that the methylene blue trihydrate intercalated into the DNA base pairs, and DNA intercalation increased the distance between DNA base pairs, depending on the types of DNA bases. According to the binding energy results, it was found that the intercalation of methylene blue trihydrate into AA-TT base pair was more favorable in the orientation of nitrogen than other directions and intercalation, and the electric charge was transferred from methylene blue trihydrate to the AA-TT base pair. The analysis of change in the charge density shows that changes often take place in the heavy atom in the middle of the system which the charge density changes most remarkable.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology