Merck
Muscarinic M3 receptors on structural cells regulate cigarette smoke-induced neutrophilic airway inflammation in mice.
American journal of physiology. Lung cellular and molecular physiology (2014-11-09)
Loes E M Kistemaker, Ronald P van Os, Albertina Dethmers-Ausema, I Sophie T Bos, Machteld N Hylkema, Maarten van den Berge, Pieter S Hiemstra, Jürgen Wess, Herman Meurs, Huib A M Kerstjens, Reinoud Gosens
ABSTRAKT
Anticholinergics, blocking the muscarinic M3 receptor, are effective bronchodilators for patients with chronic obstructive pulmonary disease. Recent evidence from M(3) receptor-deficient mice (M(3)R(-/-)) indicates that M3 receptors also regulate neutrophilic inflammation in response to cigarette smoke (CS). M(3) receptors are present on almost all cell types, and in this study we investigated the relative contribution of M(3) receptors on structural cells vs. inflammatory cells to CS-induced inflammation using bone marrow chimeric mice. Bone marrow chimeras (C56Bl/6 mice) were generated, and engraftment was confirmed after 10 wk. Thereafter, irradiated and nonirradiated control animals were exposed to CS or fresh air for four consecutive days. CS induced a significant increase in neutrophil numbers in nonirradiated and irradiated control animals (4- to 35-fold). Interestingly, wild-type animals receiving M(3)R(-/-) bone marrow showed a similar increase in neutrophil number (15-fold). In contrast, no increase in the number of neutrophils was observed in M3R(-/-) animals receiving wild-type bone marrow. The increase in keratinocyte-derived chemokine (KC) levels was similar in all smoke-exposed groups (2.5- to 5.0-fold). Microarray analysis revealed that fibrinogen-α and CD177, both involved in neutrophil migration, were downregulated in CS-exposed M(3)R(-/-) animals receiving wild-type bone marrow compared with CS-exposed wild-type animals, which was confirmed by RT-qPCR (1.6-2.5 fold). These findings indicate that the M(3) receptor on structural cells plays a proinflammatory role in CS-induced neutrophilic inflammation, whereas the M(3) receptor on inflammatory cells does not. This effect is probably not mediated via KC release, but may involve altered adhesion and transmigration of neutrophils via fibrinogen-α and CD177.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sigma-Aldrich
Fibrinogen from rat plasma, 60-80% protein (≥60% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 35-65% protein (≥90% of protein is clottable).
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)