Merck
Seminal Helicobacter pylori treatment improves sperm motility in infertile asthenozoospermic men.
Urology (2014-11-30)
Yehia El-Garem, Mohamed El-Sawy, Taymour Mostafa
ABSTRAKT
To assess the effect of treatment of seminal Helicobacter pylori in infertile asthenozoospermic men. In all, 223 infertile asthenozoospermic men were consecutively selected. They were subjected to history taking, clinical examination, semen analysis, and estimation of H pylori IgA antibodies in their seminal fluid. Infertile men with high seminal H pylori IgA were subjected to triple drug treatment, omeprazole, 20 mg; tinidazole, 500 mg; and clarithromycin, 250 mg twice a day for 2 weeks. Semen analysis as well as H pylori IgA antibodies was estimated after 3 months. In all, 22 of 223 men (9.87%) demonstrated H pylori IgA antibodies in their seminal plasma. After treatment, mean seminal H pylori IgA levels demonstrated significant decrease (1.55 ± 0.4 vs 0.52 ± 0.26; 95% confidence interval, 0.83-1.21; P = .001) concomitant with improved progressive as well as nonprogressive sperm motility. H pylori IgA antibodies demonstrated significant negative correlation with progressive sperm motility, nonprogressive sperm motility, normal sperm morphology, and significant positive correlation with immotile sperm motility. H pylori treatment significantly improves sperm motility in infertile asthenozoospermic men with elevated seminal H pylori IgA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Clarithromycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Tinidazole, VETRANAL®, analytical standard
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Omeprazole, European Pharmacopoeia (EP) Reference Standard
USP
Omeprazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Omeprazole, solid
USP
Tożsamość klarytromycyny, United States Pharmacopeia (USP) Reference Standard
Supelco
Omeprazole, analytical standard
Omeprazole for peak identification, European Pharmacopoeia (EP) Reference Standard