Merck
Thallium in spawn, juveniles, and adult common toads (Bufo bufo) living in the vicinity of a zinc-mining complex, Poland.
Environmental monitoring and assessment (2014-11-25)
Krzysztof Dmowski, Monika Rossa, Joanna Kowalska, Beata Krasnodębska-Ostręga
ABSTRAKT
A breeding population of the common toad Bufo bufo living in the vicinity of a Zn-Pb smelting works in Bukowno, Poland was studied for the presence of thallium. Tl concentration was measured in the bottom sediments of the spawning pond, in the laid eggs, in juveniles after metamorphosis, and in the selected tissues of the adult individuals. A very high concentration of Tl was detected in the spawn (13.97 ± 8.90 mg/kg d.w.). In 50% of the spawn samples, levels exceeded 20 mgTl/kg d.w. The issue of maternal transfer of thallium from females to oocytes is discussed. Due to a significant accumulation of thallium, spawn analysis can be used as a sensitive indicator of the presence of this element in the environment and may replace more invasive methods that involve the killing of adult animals. In those regions that are abundant in Zn-Pb ores, the spawn of amphibians may be a very important source of thallium contamination for predators. From among all tissues of the Bukowno adult toads, the livers have shown the highest accumulation of thallium (mean 3.98 mg/kg d.w. and maximum value--18.63). For as many as 96.5% of livers, concentrations exceeded 1.0 mgTl/kg d.w. which is treated as indicative of poisoning.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Thallium, granular, 99.9% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Zinc, foil, 1m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, wire reel, 1m, diameter 1.0mm, 99.99+%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Zinc, wire reel, 100m, diameter 1.0mm, extruded, 99.9%
Zinc, foil, 1m coil, thickness 0.125mm, as rolled, 99.95+%
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Zinc, foil, 15mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 1m coil, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 1m coil, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 15mm disks, thickness 0.005mm, 99.9%
Zinc, foil, 10mm disks, thickness 0.02mm, 99.9%
Zinc, foil, 0.5m coil, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 1.0mm, as rolled, 99.95+%