Merck
Delayed adverse reactions to the parenteral administration of iodinated contrast media.
AJR. American journal of roentgenology (2014-11-22)
Robert E Egbert, Carlo N De Cecco, U Joseph Schoepf, Andrew D McQuiston, Felix G Meinel, Richard W Katzberg
ABSTRAKT
This article presents an overview of delayed adverse reactions (DARs) to parenteral iodinated contrast media and discusses the clinical nature, risk factors, mechanisms, and potential economic implications of these DARs. DARs to contrast media are not rare but are often not recognized as being linked to contrast administration and may be falsely ascribed to other drugs. These side effects are problematic because the patient is usually without medical supervision.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%