Merck
  • Use of [(18)F]-FDG PET to predict response to neoadjuvant trastuzumab and docetaxel in patients with HER2-positive breast cancer, and addition of bevacizumab to neoadjuvant trastuzumab and docetaxel in [(18)F]-FDG PET-predicted non-responders (AVATAXHER): an open-label, randomised phase 2 trial.
Use of [(18)F]-FDG PET to predict response to neoadjuvant trastuzumab and docetaxel in patients with HER2-positive breast cancer, and addition of bevacizumab to neoadjuvant trastuzumab and docetaxel in [(18)F]-FDG PET-predicted non-responders (AVATAXHER): an open-label, randomised phase 2 trial.
The Lancet. Oncology (2014-12-03)
Bruno Coudert, Jean-Yves Pierga, Marie-Ange Mouret-Reynier, Kaldoun Kerrou, Jean-Marc Ferrero, Thierry Petit, Pierre Kerbrat, Pierre-François Dupré, Thomas Bachelot, Philippe Gabelle, Sylvia Giard, David Coeffic, Philippe Bougnoux, Jean-Briac Prevost, Gilles Paintaud, Gilles Thibault, Juana Hernandez, Mathieu Coudert, Laurent Arnould, Alina Berriolo-Riedinger
ABSTRAKT
An effective and well tolerated treatment is needed for patients with early HER2-positive breast cancer who do not achieve a pathological complete response after neoadjuvant therapy. The AVATAXHER trial aimed to predict pathological complete response early with the use of PET and to investigate whether the addition of bevacizumab could improve the proportion of patients achieving a pathological complete response in patients unlikely to respond to treatment. AVATAXHER was a randomised, open-label, non-comparative, multicentre phase 2 study that enrolled women (≥18 years of age) with early-stage HER2-positive breast cancer from 26 oncology centres in France. Patients initially received two cycles of neoadjuvant docetaxel (100 mg/m(2) intravenously every 3 weeks) plus trastuzumab (8 mg/kg intravenously every 3 weeks then 6 mg/kg intravenously every 3 weeks for the second course). Before the first and second cycles, [(18)F]-fluorodeoxyglucose (FDG) PET was done and the change in standardised uptake value was used to predict pathological complete response in each patient. Patients who were predicted to be responders on PET continued to receive standard therapy. Predicted non-responders were randomly assigned (2:1) to receive four cycles of docetaxel (100 mg/m(2) intravenously every 3 weeks) and trastuzumab (6 mg/kg intravenously every 3 weeks) plus bevacizumab (15 mg/kg intravenously every 3 weeks; group A) or continue on docetaxel plus trastuzumab alone (group B). Randomisation was open label and was done by an adaptive minimisation method. Although investigators and patients were aware of group assignment, the anatomo-pathologist in charge of centralised review of surgical samples and lymph nodes was masked to treatment assignment. The primary endpoint was centrally assessed pathological complete response according to the Chevallier classification. Efficacy analyses were done in the intention-to-treat population. Safety analyses in this Article were done on all patients who received at least one dose of treatment starting from cycle 3. Survival outcomes are not yet mature. This study is registered with ClinicalTrials.gov (NCT01142778) and EUDRACT (2009-013410-26). Between May 19, 2010, and Oct 1, 2012, 152 patients were recruited for the study. Ten patients were subsequently excluded, leaving 142 patients in the intention-to-treat population. Of these 142 patients, 69 were predicted by [(18)F]-FDG PET to be treatment responders after two cycles of treatment. The 73 predicted non-responders were randomly assigned to group A (n=48) and group B (n=25). Pathological complete responses were noted in 37 (53·6%, 95% CI 41·2-65·7) of the PET responders, 21 (43·8%, 29·5-58·8) of those in group A, and six (24·0%, 9·4-45·1) of those in group B. Incidences of grade 3-4 adverse events were similar in all three groups. The most common grade 3-4 adverse events were neutropenia (four in PET responders, five in group A, and three in group B), febrile neutropenia (one, three, and one, respectively), and myalgia (four, none, and one, respectively). Overall, 24 serious adverse events were reported in 15 patients (PET responders: nine events in four [6%] of 67 patients; group A: 14 events in ten [21%] of 47 patients; group B: one event in one [4%] of 25 patients). No deaths occurred during the study. In patients with HER2-positive breast cancer, early PET assessment can help to identify non-responders to neoadjuvant docetaxel plus trastuzumab therapy. In these patients, the addition of bevacizumab can increase the proportion of patients achieving a pathological complete response. This potential new role for PET and the activity of bevacizumab in this setting need to be confirmed in larger phase 3 trials. Roche France.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
2-Fluoro-2-deoxy-D-glucose, glycosylation inhibitor, glucose analog
Tantalum(V) ethoxide, packaged for use in deposition systems
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Anhydrous Docetaxel, European Pharmacopoeia (EP) Reference Standard
USP
Docetaksel, United States Pharmacopeia (USP) Reference Standard