Merck
Retinal output changes qualitatively with every change in ambient illuminance.
Nature neuroscience (2014-12-09)
Alexandra Tikidji-Hamburyan, Katja Reinhard, Hartwig Seitter, Anahit Hovhannisyan, Christopher A Procyk, Annette E Allen, Martin Schenk, Robert J Lucas, Thomas A Münch
ABSTRAKT
The collective activity pattern of retinal ganglion cells, the retinal code, underlies higher visual processing. How does the ambient illuminance of the visual scene influence this retinal output? We recorded from isolated mouse and pig retina and from mouse dorsal lateral geniculate nucleus in vivo at up to seven ambient light levels covering the scotopic to photopic regimes. Across each luminance transition, most ganglion cells exhibited qualitative response changes, whereas they maintained stable responses within each luminance. We commonly observed the appearance and disappearance of ON responses in OFF cells and vice versa. Such qualitative response changes occurred for a variety of stimuli, including full-field and localized contrast steps and naturalistic movies. Our results suggest that the retinal code is not fixed but varies with every change of ambient luminance. This finding raises questions about signal processing within the retina and has implications for visual processing in higher brain areas.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Supelco
γ-Aminobutyric acid, analytical standard
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
SR-95531, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat