Merck
Histone deacetylase inhibition induces long-lasting changes in maternal behavior and gene expression in female mice.
Endocrinology (2014-06-17)
Danielle S Stolzenberg, Jacqueline S Stevens, Emilie F Rissman
ABSTRAKT
In many species, including mice, maternal responsiveness is experience-dependent and permanent, lasting for long periods (months to years). We have shown that after brief exposures to pups, virgin female mice continue to respond maternally toward pups for at least one month. Administration of a histone deacetylase inhibitor (HDACi) reduces the amount of maternal experience required to affect maternal behavior and gene expression. In this set of studies, we examined the epigenetic mechanisms that underlie these motivated behaviors. We assessed whether the effects of HDACi persisted 1 month after the initial experience (in the absence of continued pup experience or HDACi treatment) and whether the maintenance of maternal memory was associated with stable changes in gene expression. Using chromatin immunoprecipitation, we examined whether Esr2 and Oxt gene expression might be mediated by recruitment of the histone acetyltransferase cAMP response element binding protein (CBP) to their promoter regions after maternal memory consolidation. We report that HDACi treatment induced long-lasting changes in maternal responsiveness. Maternal learning was associated with increased recruitment of CBP to the Esr2 and Oxt gene promoters during the consolidation of maternal memory as well as a persistent increase in estrogen receptor-β (Esr2) mRNA and decreased expression of the de novo DNA methyltransferase Dnmt3a within the medial preoptic area. The consolidation of the maternal experience may involve the CBP recruitment and stable changes in gene expression, which maintain increased maternal responsiveness for long periods of time.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium butyrate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)