Merck
Lymph node fibroblastic reticular cell transplants show robust therapeutic efficacy in high-mortality murine sepsis.
Science Translational Medicine (2014)
Fletcher AL, Elman JS, Astarita J, et al.