Merck
  • Chemistry of heterocyclic compounds. 29. Synthesis and reactions of multihetero macrocycles possessing 2, 4-pyrimidino subunits connected by carbon-oxygen and/or-sulfur linkages.
Chemistry of heterocyclic compounds. 29. Synthesis and reactions of multihetero macrocycles possessing 2, 4-pyrimidino subunits connected by carbon-oxygen and/or-sulfur linkages.
The Journal of Organic Chemistry (1978)
Newkome GR, et al.