Merck
  • Active Control of the Anchoring of 4`-Pentyl-4-cyanobiphenyl (5CB) at an Aqueous?Liquid Crystal Interface By Using a Redox-Active Ferrocenyl Surfactant
Active Control of the Anchoring of 4`-Pentyl-4-cyanobiphenyl (5CB) at an Aqueous?Liquid Crystal Interface By Using a Redox-Active Ferrocenyl Surfactant
Langmuir
Brake JM, et al.