Merck
1-Arylsulfonyl-2-(pyridylmethylsulfinyl) benzimidazoles as new proton pump inhibitor prodrugs.
Molecules (Basel, Switzerland) (2009-12-25)
Jai Moo Shin, George Sachs, Young-moon Cho, Michael Garst
ABSTRAKT
New arylsulfonyl proton pump inhibitor (PPI) prodrug forms were synthesized. These prodrugs provided longer residence time of an effective PPI plasma concentration, resulting in better gastric acid inhibition.