Merck
Rif1 maintains telomere length homeostasis of ESCs by mediating heterochromatin silencing.
Developmental cell (2014-04-17)
Jiameng Dan, Yifei Liu, Na Liu, Maria Chiourea, Maja Okuka, Tao Wu, Xiaoying Ye, Chunlin Mou, Lei Wang, Lingling Wang, Yu Yin, Jihong Yuan, Bingfeng Zuo, Fang Wang, Zhiguo Li, Xinghua Pan, Zhinan Yin, Lingyi Chen, David L Keefe, Sarantis Gagos, Andrew Xiao, Lin Liu
ABSTRAKT
Telomere length homeostasis is essential for genomic stability and unlimited self-renewal of embryonic stem cells (ESCs). We show that telomere-associated protein Rif1 is required to maintain telomere length homeostasis by negatively regulating Zscan4 expression, a critical factor for telomere elongation by recombination. Depletion of Rif1 results in terminal hyperrecombination, telomere length heterogeneity, and chromosomal fusions. Reduction of Zscan4 by shRNA significantly rescues telomere recombination defects of Rif1-depleted ESCs and associated embryonic lethality. Further, Rif1 negatively modulates Zscan4 expression by maintaining H3K9me3 levels at subtelomeric regions. Mechanistically, Rif1 interacts and stabilizes H3K9 methylation complex. Thus, Rif1 regulates telomere length homeostasis of ESCs by mediating heterochromatic silencing.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało przeciw trimetylo-histonowi H3 (Lys9), Upstate®, from rabbit