Merck
<BIG>Competitive flow immunoassay with fluorescence detection for determination of 4-nitrophenol.</BIG>
Analytica Chimica Acta (2001)
<BIG>Nistor, C., Oubina, A., Marco, M. P., Barcelo, D., &amp; Emneus, J.</BIG>