Merck
GalNAc-T14 may be involved in regulating the apoptotic action of IGFBP-3.
Journal of biosciences (2009-10-07)
Chen Wu, Yaojun Shan, Xinxia Liu, Wenqian Song, Jiali Wang, Minji Zou, Min Wang, Donggang Xu
ABSTRAKT
Insulin-like growth factor binding protein-3 (IGFBP-3) is known to induce apoptosis in an insulin-like growth factor (IGF)-dependent and IGF-independent manner, but the mechanism underlying the IGF-independent effects remains unclear. Polypeptide N-acetylgalactosaminyltransferase 14 (GalNAc-T14) is a novel IGFBP-3 binding partner. In this paper, small interference RNA (siRNA) targeting GalNAc-T14 was used to examine whether GalNAc-T14 affects the apoptotic action of IGFBP-3. Using semi-quantitative reverse-transcriptase polymerase chain reaction (RT-PCR) and western blot analysis, we determined that GalNAc-T14 expression was downregulated by the siRNA directed against GalNAc-T14. Apoptosis analysis of IGFBP-3-overexpressing cells treated with siRNA against GalNAc-T14 was performed to determine if GalNAc-T14 was specifically involved in IGFBP-3 signalling. The results, as determined by flow cytometric analysis and caspase-3 assay, showed that the extent of apoptosis induced by IGFBP-increased with RNA interference (RNAi) knockdown of GalNAc-T14. Our data suggest that GalNAc-T14 influences the apoptotic action of IGFBP-3 and might mediate the signalling pathway of IGFBP-3. Experiments to determine the role of GalNAc-T14 in the regulation of apoptosis induced by IGFBP-3 are under way.