Merck
Molecular characterization of a novel short-chain dehydrogenase/reductase that reduces all-trans-retinal.
The Journal of biological chemistry (1998-08-15)
F Haeseleer, J Huang, L Lebioda, J C Saari, K Palczewski
ABSTRAKT
The reduction of all-trans-retinal in photoreceptor outer segments is the first step in the regeneration of bleached visual pigments. We report here the cloning of a dehydrogenase, retSDR1, that belongs to the short-chain dehydrogenase/reductase superfamily and localizes predominantly in cone photoreceptors. retSDR1 expressed in insect cells displayed substrate specificities of the photoreceptor all-trans-retinol dehydrogenase. Homology modeling of retSDR1 using the carbonyl reductase structure as a scaffold predicted a classical Rossmann fold for the nucleotide binding, and an N-terminal extension that could facilitate binding of the enzyme to the cell membranes. The presence of retSDR1 in a subset of inner retinal neurons and in other tissues suggests that the enzyme may also be involved in retinol metabolism outside of photoreceptors.