Merck
  • Arginine transport is impaired in C57Bl/6 mouse macrophages as a result of a deletion in the promoter of Slc7a2 (CAT2), and susceptibility to Leishmania infection is reduced.
Arginine transport is impaired in C57Bl/6 mouse macrophages as a result of a deletion in the promoter of Slc7a2 (CAT2), and susceptibility to Leishmania infection is reduced.
The Journal of infectious diseases (2013-03-06)
M Gloria Sans-Fons, Andrée Yeramian, Selma Pereira-Lopes, Luis F Santamaría-Babi, Manuel Modolell, Jorge Lloberas, Antonio Celada
ABSTRAKT
Host genetic factors play a crucial role in immune response. To determine whether the differences between C57Bl/6 and BALB-C mice are due only to the production of cytokines by T-helper 1 cells or T-helper 2 cells, we obtained bone marrow-derived macrophages from both strains and incubated them with these cytokines. Although the induction of Nos2 and Arg1 was similar in the 2 strains, infectivity to Leishmania major differed, as did macrophage uptake of arginine, which was higher in BALB-C macrophages. The levels of interferon γ- and interleukin 4-dependent induction of the cationic amino acid transporter SLC7A2 (also known as "cationic amino acid transporter 2," or "CAT2") were decreased in macrophages from C57Bl/6 mice. This reduction was a result of a deletion in the promoter of one of the 4 AGGG repeats. These results demonstrate that the availability of arginine controls critical aspects of macrophage activation and reveal a factor for susceptibility to Leishmania infection.