Merck
Roles of CUB domain-containing protein 1 signaling in cancer invasion and metastasis.
Cancer science (2011-08-05)
Takamasa Uekita, Ryuichi Sakai
ABSTRAKT
Tumor metastasis is a complex multistep process by which cells from the primary tumor invade tissues, move through the vasculature, settle at distant sites and eventually grow to form secondary tumors. Altered tyrosine phosphorylation signals in cancer cells contribute to a number of aberrant characteristics involved in tumor invasion and metastasis. CUB domain-containing protein 1 (CDCP1) is a substrate of Src family kinases and has been shown to regulate anoikis resistance, migration and matrix degradation during tumor invasion and metastasis in a tyrosine phosphorylation-dependent manner. Knockdown of CDCP1 blocks tumor metastasis or peritoneal dissemination in vivo, without significantly affecting cell proliferation. Moreover, expression levels of CDCP1 are of prognostic value in several cancers. Here, we summarize the studies on CDCP1, focusing on structure and signal transduction, to gain insight into its role in cancer progression. Understanding the signaling pathways regulated by CDCP1 could help establish novel therapeutic strategies against the progression of cancer.