Merck
Expression of the human cancer/testis antigen NY-SAR-35 is activated by CpG island hypomethylation.
Biotechnology letters (2011-02-15)
Jin-Hyung Park, Myung-Ha Song, Chang-Hun Lee, Min-Ki Lee, Young-Min Park, Lloyd Old, Sang-Yull Lee
ABSTRAKT
The novel cancer/testis antigen gene, NY-SAR-35, is expressed exclusively in normal testis and in various histological types of tumor. However, the NY-SAR-35 gene expression is observed to be aberrant in several cancer cell lines and tissues. The analysis of methylation status of the NY-SAR-35 gene promoter in various cancer cell lines showed that its expression was related to methylation of the promoter region. Treatment of human cancer cell lines with the demethylating agent 5-aza-2'-deoxycytidine activated the expression of the NY-SAR-35 gene. In addition, transfection experiments on various fragments of the CpG-rich gene promoter indicate that in vitro methylation of the NY-SAR-35 gene promoter results in the loss of promoter activity. The expression of NY-SAR-35 is therefore activated by hypomethylation of the CpG island in the gene promoter.