Merck
  • The antibacterial effect of camphorated paramonochlorophenol, camphorated phenol and calcium hydroxide in the treatment of infected root canals.
The antibacterial effect of camphorated paramonochlorophenol, camphorated phenol and calcium hydroxide in the treatment of infected root canals.
Endodontics & dental traumatology (1985-10-01)
A Bystrom, R Claesson, G Sundqvist
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium hydroxide, ACS reagent, ≥95.0%