- Enzymatic chemiluminescence assay for inorganic pyrophosphate.
Enzymatic chemiluminescence assay for inorganic pyrophosphate.
Analytical biochemistry (2002-04-24)
Vuokko Jansson, Kristian Jansson
PMID11969200
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyrophosphatase, Inorganic from baker′s yeast (S. cerevisiae), powder, ≥500 units/mg protein (E1%/280)