Merck
  • Influence of mild hypothermia on inducible nitric oxide synthase expression and reactive nitrogen production in experimental stroke and inflammation.
Influence of mild hypothermia on inducible nitric oxide synthase expression and reactive nitrogen production in experimental stroke and inflammation.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2002-05-23)
Hyung Soo Han, Yanli Qiao, Murat Karabiyikoglu, Rona G Giffard, Midori A Yenari
ABSTRAKT
Mild hypothermia is neuroprotective, but the reasons are not well known. Inflammation contributes to ischemic damage; therefore, we examined whether the protection by hypothermia may be attributable to alterations in the inflammation. We examined whether hypothermia might alter the inflammatory cell-associated inducible nitric oxide synthase (iNOS) and subsequent nitric oxide (NO) and peroxynitrite generation in experimental stroke and inflammation. Rats underwent 2 hr of middle cerebral artery occlusion (MCAO). Brain inflammation was modeled by intravenous lipopolysaccharide (LPS) (2 mg/kg) injection. Temperature was maintained at 33 degrees C for 2 hr immediately after MCAO and LPS injection, delayed 2 hr after MCAO or maintained at 38 degrees C. Cultured microglia were activated with LPS and then incubated at 33 or 37 degrees C. Both intraischemic and delayed mild hypothermia attenuated infarct size by 40% (p < 0.05). Immunohistochemistry was performed to identify cell type, iNOS, and peroxynitrite. The majority of iNOS- and peroxynitrite-positive cells were activated microglia-macrophages, and mild hypothermia significantly decreased the numbers of immunoreactive cells at 72 hr by >50% (p < 0.05). After ischemia, mild hypothermia decreased NO production by 40%. Similarly, hypothermia attenuated NO and iNOS in LPS-injected rats, as well as in cultured microglia. Aminoguanidine, an iNOS inhibitor, also attenuated infarct size and NO in ischemic and inflammation models. We conclude that mild hypothermia significantly inhibits the inflammatory response by affecting microglial iNOS-NO generation. Therapies directed against microglia or their activation may be useful in treating stroke.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Griess reagent (modified)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, ascites fluid, clone HM-2
Sigma-Aldrich
Anti-Nitric Oxide Synthase (1131-1144), Inducible Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Przeciwciało przeciw nitrotyrozynie, Upstate®, from rabbit