Merck
Androgen receptor controls EGFR and ERBB2 gene expression at different levels in prostate cancer cell lines.
Cancer research (2009-03-26)
Jean-Christophe Pignon, Benjamin Koopmansch, Gregory Nolens, Laurence Delacroix, David Waltregny, Rosita Winkler
ABSTRAKT
EGFR or ERBB2 contributes to prostate cancer (PCa) progression by activating the androgen receptor (AR) in hormone-poor conditions. Here, we investigated the mechanisms by which androgens regulate EGFR and ERBB2 expression in PCa cells. In steroid-depleted medium (SDM), EGFR protein was less abundant in androgen-sensitive LNCaP than in androgen ablation-resistant 22Rv1 cells, whereas transcript levels were similar. Dihydrotestosterone (DHT) treatment increased both EGFR mRNA and protein levels and stimulated RNA polymerase II recruitment to the EGFR gene promoter, whereas it decreased ERBB2 transcript and protein levels in LNCaP cells. DHT altered neither EGFR or ERBB2 levels nor the abundance of prostate-specific antigen (PSA), TMEPA1, or TMPRSS2 mRNAs in 22Rv1 cells, which express the full-length and a shorter AR isoform deleted from the COOH-terminal domain (ARDeltaCTD). The contribution of both AR isoforms to the expression of these genes was assessed by small interfering RNAs targeting only the full-length or both AR isoforms. Silencing of both isoforms strongly reduced PSA, TMEPA1, and TMPRSS2 transcript levels. Inhibition of both AR isoforms did not affect EGFR and ERBB2 transcript levels but decreased EGFR and increased ERBB2 protein levels. Proliferation of 22Rv1 cells in SDM was inhibited in the absence of AR and ARDeltaCTD. A further decrease was obtained with PKI166, an EGFR/ERBB2 kinase inhibitor. Overall, we showed that ARDeltaCTD is responsible for constitutive EGFR expression and ERBB2 repression in 22Rv1 cells and that ARDeltaCTD and tyrosine kinase receptors are necessary for sustained 22Rv1 cell growth.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Roche
X-tremeGENE siRNA Transfection Reagent, Polymer reagent for delivering siRNA to common cell lines