Merck
  • Biosynthetic origin of conjugated double bonds: production of fatty acid components of high-value drying oils in transgenic soybean embryos.
Biosynthetic origin of conjugated double bonds: production of fatty acid components of high-value drying oils in transgenic soybean embryos.
Proceedings of the National Academy of Sciences of the USA (1999)
Cahoon EB, et al.