Merck
Safety and immunogenicity of an MF59-adjuvanted A/H1N1 pandemic influenza vaccine in children from three to seventeen years of age.
Vaccine (2014-12-03)
Markus Knuf, Geert Leroux-Roels, Hans C Rümke, Katia Abarca, Luis Rivera, Maria Lattanzi, Paola Pedotti, Ashwani Arora, Dorothee Kieninger-Baum, Giovanni Della Cioppa
ABSTRAKT
This study was designed to identify the optimal dose of an MF59-adjuvanted, monovalent, A/H1N1 influenza vaccine in healthy paediatric subjects. Subjects aged 3-8 years (n=194) and 9-17 years (n=160) were randomized to receive two primary doses of A/H1N1 vaccine containing either 3.75 μg antigen with half a standard dose of MF59 adjuvant, 7.5 μg antigen with a full dose of MF59, or (children 3-8 years only), a non-adjuvanted 15 μg formulation. A booster dose of MF59-adjuvanted seasonal influenza vaccine including homologous A/H1N1 strain was given one year after priming. Immunogenicity was assessed by haemagglutination inhibition (HI) and microneutralization assays. Vaccine safety was assessed throughout the study (up to 18 months). A single priming dose of either MF59-adjuvanted formulation was sufficient to meet the European licensure criteria for pandemic influenza vaccines (HI titres ≥1:40>70%; seroconversion>40%; and GMR>2.5). Two non-adjuvanted vaccine doses were required to meet the same licensure criteria. After first and second doses, percentage of subjects with HI titres ≥1:40 were between 97% and 100% in the adjuvanted vaccine groups compared with 68% and 91% in the non-adjuvanted group, respectively. Postvaccination seroconversion rates ranged from 91% to 98% in adjuvanted groups and were 68% (first dose) and 98% (second dose) in the non-adjuvanted group. HI titres ≥1:330 after primary doses were achieved in 69% to 90% in adjuvanted groups compared with 41% in the non-adjuvanted group. Long-term antibody persistence after priming and a robust antibody response to booster immunization were observed in all vaccination groups. All A/H1N1 vaccine formulations were generally well tolerated. No vaccine-related serious adverse events occurred, and no subjects were withdrawn from the study due to an adverse event. An MF59-adjuvanted influenza vaccine containing 3.75 μg of A/H1N1 antigen was well tolerated and sufficiently immunogenic to meet all the European licensure criteria after a single dose in healthy children 3-17 years old.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Squalene, analytical standard
Sigma-Aldrich
Squalene, ≥98%, liquid