Merck
Optomotor-blind negatively regulates Drosophila eye development by blocking Jak/STAT signaling.
PloS one (2015-03-18)
Yu-Chen Tsai, Stefan Grimm, Ju-Lan Chao, Shih-Chin Wang, Kerstin Hofmeyer, Jie Shen, Fred Eichinger, Theoni Michalopoulou, Chi-Kuang Yao, Chih-Hsuan Chang, Shih-Han Lin, Y Henry Sun, Gert O Pflugfelder
ABSTRAKT
Organ formation requires a delicate balance of positive and negative regulators. In Drosophila eye development, wingless (wg) is expressed at the lateral margins of the eye disc and serves to block retinal development. The T-box gene optomotor-blind (omb) is expressed in a similar pattern and is regulated by Wg. Omb mediates part of Wg activity in blocking eye development. Omb exerts its function primarily by blocking cell proliferation. These effects occur predominantly in the ventral margin. Our results suggest that the primary effect of Omb is the blocking of Jak/STAT signaling by repressing transcription of upd which encodes the Jak receptor ligand Unpaired.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phosphorus, red, ≥97.0%
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis