Merck
A (99m)Tc-labeled misonidazole analogue: step toward a (99m)Tc-alternative to [18F]fluromisonidazole for detecting tumor hypoxia.
Cancer biotherapy & radiopharmaceuticals (2015-01-09)
Madhava B Mallia, Anupam Mathur, Haladhar D Sarma, Sharmila Banerjee
ABSTRAKT
The PET radiopharmaceutical [(18)F]Fluromisonidazole ([(18)F]FMISO) is presently the agent of choice for the clinical imaging of tumor hypoxia. Considering the logistic advantages of (99m)Tc and wider availability of SPECT machines, a (99m)Tc-radiopharmaceutical for this purpose constitutes an attractive choice. In the work presented here, a misonidazole analogue was radiolabeled with (99m)Tc(CO)3 core and the complex was evaluated in Swiss mice bearing fibrosarcoma tumor. The results obtained are compared with the biodistribution of [(18)F]FMISO carried out in the same tumor-bearing animal model. Misonidazole-(99m)Tc(CO)3 complex showed significant uptake and retention in tumor. Notably, the rate of clearance of misonidazole complex from the tumor was slower than that of [(18)F]FMISO. The maximum tumor/muscle ratio obtained with misonidazole-(99m)Tc(CO)3 complex was significantly higher than that of [(18)F]FMISO. The study constitutes a positive step toward the development of a (99m)Tc-analogue of [(18)F]FMISO.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Nitroimidazole, 98%
Sigma-Aldrich
N-Ethyldiisopropylamine, ≥98.0%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
tert-Butyl bromoacetate, 98%