Merck
The multi-targets integrated fingerprinting for screening anti-diabetic compounds from a Chinese medicine Jinqi Jiangtang Tablet.
Journal of ethnopharmacology (2015-02-24)
Yan-Xu Chang, Ai-Hua Ge, Sineeporn Donnapee, Jin Li, Yang Bai, Jiao Liu, Jun He, Xi Yang, Li-Jiao Song, Bo-Li Zhang, Xiu-Mei Gao
ABSTRAKT
Jinqi Jiangtang Tablet is a traditional Chinese anti-diabetic formula containing three ingredients: Coptis chinensis Franch. (dried rhizome of C. chinensis Franch., Coptis deltoidea C. Y. Cheng et Hsiao and Coptis teeta Wall.), Astragalus membranaceus (Fisch.) Bunge. (dried root of A. membranaceus (Fisch.) Bge. var. mongholicus (Bge. ) Hsiao and A. membranaceus (Fisch.) Bge. ) and Lonicera japonica Thunb. (dried alabastrum or with nascent flowers of L. japonica Thunb. ). Free radicals, α-glucosidase, α-amylase, aldose reductase and lipase are different targets related with diabetes. However, there are no chromatographic methods employed in screening the anti-diabetic compounds from natural products basing on these targets simultaneously. The present study was aimed at the establishment of a multi-targets integrated fingerprinting to clarify the possible mechanism of the action of Traditional Chinese Medicines which simultaneously contained multiple chemical characteristics and effects of constitutions. The multi-targets integrated fingerprinting was developed and validated to screen anti-diabetic compounds from natural products by using ultra-high-performance liquid chromatography/quadruple-time-of-flight mass spectrometry, fraction collector and microplate reader. Ultra performance liquid chromatography was employed to separate the components in Jinqi Jiangtang Tablet, which were identified by quadruple-time-of-flight mass spectrometry to acquire their structural information and collected by the fraction collector. Finally the active fractions were tested for scavenging 1, 1-diphenyl-2-picrylhydrazyl radical and inhibition of α-glucosidase, α-amylase, aldose reductase, and lipase activities in vitro by microplate reader. Our tests revealed that the Jinqi Jiangtang Tablet showed inhibitory activity against α-glucosidase, α-amylase, aldose reductase and lipase with IC50 values of 0.80 ± 0.02 mg/mL, 1.28 ± 0.13 mg/mL, 0.80 ± 0.02 mg/mL, 1.90 ± 0.18 mg/mL respectively and the scavenging activity with IC50 value of 1.71 ± 0.178 mg/mL. The bioactive fractions were identified to be alkaloids, flavonoids and phenolic acids. The phenolic acids possessed antioxidant activities, namely the scavenging effect on 1, 1-diphenyl-2-picrylhydrazyl rull;). The alkaloids exhibited inhibitory activity against α-glucosidase, aldose reductase, α-amylase, and lipase. The flavonoids also showed mild inhibition on α-glucosidase, aldose reductase, α-amylase and lipase. The results demonstrate that Jinqi Jiangtang Tablet can scavenge free radicals and inhibit α-glucosidase, aldose reductase, α-amylase and lipase, which may be the possible mechanism of action of Jinqi Jiangtang Tablet for the treatment of diabetes and associated complications. Compared with conventional chromatographic separation and activity assays, the multi-targets integrated fingerprinting, which simultaneously contains the chemical characteristics and multiple effects of constitutions could comprehensively and properly reveal the activity of Jinqi Jiangtang Tablet. The results also show that the multi-targets integrated fingerprinting is a novel and powerful tool for screening and identifying active ingredients from Traditional Chinese Medicines.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Nitrophenyl β-D-glucuronide, ≥98% (TLC)
Sigma-Aldrich
4-Nitrophenyl β-D-glucuronide, ≥99.0% (TLC)
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%