Merck
VCP/Cdc48 rescues the growth defect of a GPI10 mutant in yeast.
FEBS letters (2015-01-28)
Yohei Ohnuma, Takahiro Takata, Junko Kawawaki, Kunihiko Yasuda, Keiji Tanaka, Yoko Kimura, Akira Kakizuka
ABSTRAKT
We identified a yeast mutant with temperature-sensitive growth defects that were rescued by VCP expression. The mutation occurred in GPI10, which encodes a mannosyl transferase for glycosylphosphatidylinositol anchor formation in the endoplasmic reticulum, and caused a Gly469Glu substitution in Gpi10. The mutant exhibited increased unfolded protein response, which was partially rescued by VCP or Cdc48, and showed sensitivity against cell-wall stressors, which were not rescued by VCP. These results suggest a potential link between VCP/Cdc48 and Gpi10 functions in the control of cell growth.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ethyl methanesulfonate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethyl methanesulfonate, liquid