Merck
Genomic locus on chromosome 1 regulates susceptibility to spontaneous arthritis in mice deficiency of IL-1RA.
BMC immunology (2014-12-10)
Nan Deng, Yan Jiao, Yanhong Cao, Xiaoyun Liu, Yonghui Ma, Karen A Hasty, David D Brand, John M Stuart, Weikuan Gu
ABSTRAKT
To understand the role of genetic factors on chromosome 1 in the regulation of spontaneous arthritis in mice deficient in IL-1 receptor antagonist protein (IL_1RA), we previously used speed congenic breeding to transfer the QTL region from DBA/1(-/-) mice that are resistant to spontaneous arthritis into BALB/c(-/-) mice which are susceptible. We were able to establish two congenic strains which exhibited a delayed onset and reduced severity of disease. In this study, we asked a different set of questions. How will the QTL region from BALB/c(-/-) interact with the rest of the genome in the DBA/1(-/-) background? Will the DBA/1(-/-) mice become susceptible to spontaneous arthritis if the QTL genomic region on chromosome 1 was replaced with the genomic fragment of the same region from BALB/c(-/-)? We conducted the congenic breeding with the similar procedure as that of congenic strains with BALB/c(-/-) background. Instead of BALB/c(-/-), DBA/1(-/-) was used as the recurrent parent while BALB/c(-/-) was used as the donor parent. By the 6(th) generation we determined that all of the chromosomes in the progeny were of DBA/1(-/-) origin with the exception of the QTL portion of chromosome 1 which is heterozygous of BALB/c(-/-) and DBA/1(-/-) origin. We then intercrossed selected mice to produce homozygous strains containing the homozygous genomic region of BALB/c(-/-) on chromosome 1, while the rest of genome are homozygous DBA/1(-/-). This strain was observed for the development of spontaneous arthritis. Up to 9 weeks of age, both congenic strain and DBA/1(-/-) did not develop arthritis. However, after 9 weeks, the congenic strain started to exhibit signs of arthritis, while the DBA/1(-/-) remained free from disease. The result indicates a strong influence of genetic factor(s) on the QTL of chromosome 1 on the susceptibility to spontaneous arthritis. Identification of genetic factors within this QTL region in the future will significantly enhance our understanding of molecular mechanism of spontaneous arthritis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Propanediol, 98%