Merck
A stretchable nanowire UV-Vis-NIR photodetector with high performance.
Advanced materials (Deerfield Beach, Fla.) (2015-01-24)
Jewon Yoo, Sanghwa Jeong, Sungjee Kim, Jung Ho Je
ABSTRAKT
A simple direct-writing technique can be used to fabricate a stretchable UV-vis-NIR nanowire photodetector (NWPD) consisting of PbS quantum dot (QD)-poly(3-hexylthiopehene) (P3HT) hybrid NWs. The hybrid NWPD shows superior sensitivity and response speed in the UV-vis to NIR range. The stretchable UV-vis-NIR NWPD shows a nearly identical photoresponse under extreme (up to 100%) and repeated (up to 100 cycles) stretching conditions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trioctylphosphine, technical grade, 90%
Sigma-Aldrich
Trioctylphosphine, 97%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
Hexamethyldisilathiane, synthesis grade
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Supelco
Oleic acid, Selectophore, ≥99%
Supelco
1-Octadecene, analytical standard, ≥99.0% (GC)
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Oleic acid, analytical standard