Merck
KSR-based medium improves the generation of high-quality mouse iPS cells.
PloS one (2014-08-30)
Kai Liu, Fang Wang, Xiaoying Ye, Lingling Wang, Jiao Yang, Jingzhuo Zhang, Lin Liu
ABSTRAKT
Induced pluripotent stem (iPS) cells from somatic cells have great potential for regenerative medicine. The efficiency in generation of iPS cells has been significantly improved in recent years. However, the generation of high-quality iPS cells remains of high interest. Consistently, we demonstrate that knockout serum replacement (KSR)-based medium accelerates iPS cell induction and improves the quality of iPS cells, as confirmed by generation of chimeras and all iPS cell-derived offspring with germline transmission competency. Both alkaline phosphatase (AP) activity assay and expression of Nanog have been used to evaluate the efficiency of iPS cell induction and formation of ES/iPS cell colonies; however, appropriate expression of Nanog frequently indicates the quality of ES/iPS cells. Interestingly, whereas foetal bovine serum (FBS)-based media increase iPS cell colony formation, as revealed by AP activity, KSR-based media increase the frequency of iPS cell colony formation with Nanog expression. Furthermore, inhibition of MAPK/ERK by a specific inhibitor, PD0325901, in KSR- but not in FBS-based media significantly increases Nanog-GFP+ iPS cells. In contrast, addition of bFGF in KSR-based media decreases proportion of Nanog-GFP+ iPS cells. Remarkably, PD can rescue Nanog-GFP+ deficiency caused by bFGF. These data suggest that MAPK/ERK pathway influences high quality mouse iPS cells and that KSR- and PD-based media could enrich homogeneous authentic pluripotent stem cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-1 Antibody, clone MC-480, clone MC-480, Chemicon®, from mouse
Supelco
Sodium butyrate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)