Merck
  • Lonidamine induces intracellular tumor acidification and ATP depletion in breast, prostate and ovarian cancer xenografts and potentiates response to doxorubicin.
Lonidamine induces intracellular tumor acidification and ATP depletion in breast, prostate and ovarian cancer xenografts and potentiates response to doxorubicin.
NMR in biomedicine (2014-12-17)
Kavindra Nath, David S Nelson, Daniel F Heitjan, Dennis B Leeper, Rong Zhou, Jerry D Glickson
ABSTRAKT
We demonstrate that the effects of lonidamine (LND, 100 mg/kg, i.p.) are similar for a number of xenograft models of human cancer including DB-1 melanoma and HCC1806 breast, BT-474 breast, LNCaP prostate and A2870 ovarian carcinomas. Following treatment with LND, each of these tumors exhibits a rapid decrease in intracellular pH, a small decrease in extracellular pH, a concomitant monotonic decrease in nucleoside triphosphate and an increase in inorganic phosphate over a 2-3 h period. We have previously demonstrated that selective intracellular tumor acidification potentiates response of this melanoma model to melphalan (7.5 mg/kg, i.v.), producing an estimated 89% cell kill based on tumor growth delay analysis. We now show that, in both DB-1 melanoma and HCC1806 breast carcinoma, LND potentiates response to doxorubicin, producing 95% cell kill in DB-1 melanoma at 7.5 mg/kg, i.v. doxorubicin and 98% cell kill at 10.0 mg/kg doxorubicin, and producing a 95% cell kill in HCC1806 breast carcinoma at 12.0 mg/kg doxorubicin. Potentiation of doxorubicin may result from cation trapping of the weakly basic anthracycline. Recent experience with the clinical treatment of melanoma and other forms of human cancer suggests that these diseases will probably not be cured by a single therapeutic procedure other than surgery. A multimodality therapeutic approach will be required. As a potent modulator of tumor response to N-mustards and anthracyclines as well as tumor thermo- and radiosensitivity, LND promises to play an important clinical role in the management and possible complete local control of a number of prevalent forms of human cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS