Merck
  • Quantitative study of parathyroid hormone (1-34) and bone morphogenetic protein-2 on spinal fusion outcomes in a rabbit model of lumbar dorsolateral intertransverse process arthrodesis.
Quantitative study of parathyroid hormone (1-34) and bone morphogenetic protein-2 on spinal fusion outcomes in a rabbit model of lumbar dorsolateral intertransverse process arthrodesis.
Spine (2013-12-25)
Ioan A Lina, Varun Puvanesarajah, Jason A Liauw, Sheng-fu L Lo, David R Santiago-Dieppa, Lee Hwang, Annie Mao, Ali Bydon, Jean-Paul Wolinsky, Daniel M Sciubba, Ziya Gokaslan, Christina Holmes, Timothy F Witham
ABSTRAKT
A posterolateral rabbit spinal fusion model was used to evaluate the effects of recombinant human bone morphogenetic protein-2 (rhBMP-2) and teriparatide (PTH [1-34]) used individually and in combination on spinal fusion outcomes. To test the efficacy of parathyroid hormone on improving spinal fusion outcomes when used with BMP-2. Of the more than 250,000 spinal fusion surgical procedures performed each year, 5% to 35% of these will result in pseudarthrosis. Growing controversy on the efficacy and cost of rhBMP-2 for improving spinal fusion outcomes has presented a challenge for clinicians. Research into PTH as an adjunct therapy to rhBMP-2 for spinal fusion has not yet been investigated. Forty-eight male New Zealand white rabbits underwent bilateral posterolateral intertransverse process arthrodesis surgery at the L5-L6 level. Animals were divided into 6 groups. Two groups were treated with autograft alone or autograft and PTH (1-34), whereas the other 4 groups were treated with low-dose rhBMP-2 alone, high-dose rhBMP-2 alone, or either dose combined with PTH (1-34). All animals were euthanized 6 weeks after surgery. The L4-L7 spinal segment was removed and assessed using manual palpation, computed tomography (CT), and biomechanical testing. CT assessments revealed fusion in 50% of autograft controls, 75% of autograft PTH (1-34) animals, 87.5% in the 2 groups treated with low-dose rhBMP-2, and 100% in the 2 groups treated with high-dose rhBMP-2. CT volumetric analysis demonstrated that all groups treated with biologics had fusion masses that were on average significantly larger than those observed in the control group (P < 0.0001). Biomechanical data demonstrated no statistical difference between controls, PTH (1-34), and low-dose rhBMP-2 in any testing orientation. PTH (1-34) did not increase bending stiffness when used adjunctively with either low-dose or high-dose rhBMP-2. Although intermittent teriparatide administration results in increased fusion mass volume, it does not improve biomechnical stiffness over use of autograft alone. When delivered concurrently with high- and low-dose rhBMP-2, teriparatide provided no statistically significant improvement in biomechanical stiffness. N/A.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Calcium phosphate tribasic, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Hydroxyapatite, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic