Merck
DNA barcoding reveals diverse growth kinetics of human breast tumour subclones in serially passaged xenografts.
Nature communications (2014-12-24)
Long V Nguyen, Claire L Cox, Peter Eirew, David J H F Knapp, Davide Pellacani, Nagarajan Kannan, Annaick Carles, Michelle Moksa, Sneha Balani, Sohrab Shah, Martin Hirst, Samuel Aparicio, Connie J Eaves
ABSTRAKT
Genomic and phenotypic analyses indicate extensive intra- as well as intertumoral heterogeneity in primary human malignant cell populations despite their clonal origin. Cellular DNA barcoding offers a powerful and unbiased alternative to track the number and size of multiple subclones within a single human tumour xenograft and their response to continued in vivo passaging. Using this approach we find clone-initiating cell frequencies that vary from ~1/10 to ~1/10,000 cells transplanted for two human breast cancer cell lines and breast cancer xenografts derived from three different patients. For the cell lines, these frequencies are negatively affected in transplants of more than 20,000 cells. Serial transplants reveal five clonal growth patterns (unchanging, expanding, diminishing, fluctuating or of delayed onset), whose predominance is highly variable both between and within original samples. This study thus demonstrates the high growth potential and diverse growth properties of xenografted human breast cancer cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Butyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
USP
Butylparaben, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)