Merck
Circulating adipocyte fatty acid binding protein is increased in chronic and acute renal dysfunction.
Nutrition, metabolism, and cardiovascular diseases : NMCD (2014-05-13)
T Ebert, L M Hopf, U Wurst, A Bachmann, S Kralisch, U Lössner, M Platz, J Kratzsch, J U Stolzenburg, A Dietel, O Grisk, J Beige, M Anders, I Bast, N Klöting, M Blüher, M Stumvoll, M Fasshauer
ABSTRAKT
The adipokine adipocyte fatty acid binding protein (AFABP) is positively associated with the development of the metabolic syndrome, diabetes mellitus, and cardiovascular disease. We hypothesized that AFABP also increases with deteriorating renal function. Serum AFABP levels were quantified by enzyme linked immunosorbent assay in 532 patients with chronic kidney disease (CKD) covering the whole spectrum of estimated glomerular filtration rate (eGFR) categories from G1 to G5 (study population 1). Furthermore, AFABP was measured in 32 patients before and within 30 h after elective unilateral nephrectomy, a model of acute kidney dysfunction (AKD) (study population 2). Moreover, circulating AFABP was investigated in rats undergoing bilateral nephrectomy (BNE) as compared to sham-operated animals. Median serum AFABP levels adjusted for age, gender, and body mass index significantly increased with increasing eGFR category (G1: 22.0 μg/l; G2: 34.6 μg/l; G3: 56.7 μg/l; G4: 95.2 μg/l; and G5: 173.9 μg/l). Furthermore, renal dysfunction remained positively associated with AFABP in multivariate analysis in this cohort. In patients undergoing unilateral nephrectomy, AFABP increased significantly after surgery (42.1 μg/l) as compared to pre-surgical values (29.3 μg/l). Furthermore, relative changes of post-to-pre-surgical AFABP levels were independently associated with relative changes of post-to-pre-surgical creatinine concentrations. After BNE in rats, AFABP increased significantly as compared to sham-operated animals. We show that AFABP is significantly elevated in CKD and AKD patients. Furthermore, measures of renal function are associated with circulating AFABP. Moreover, animal experiments indicate that AFABP levels strongly depend on renal function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material