Merck
Phosphorus sequestration in the form of polyphosphate by microbial symbionts in marine sponges.
Proceedings of the National Academy of Sciences of the United States of America (2015-02-26)
Fan Zhang, Leah C Blasiak, Jan O Karolin, Ryan J Powell, Chris D Geddes, Russell T Hill
ABSTRAKT
Marine sponges are major habitat-forming organisms in coastal benthic communities and have an ancient origin in evolution history. Here, we report significant accumulation of polyphosphate (polyP) granules in three common sponge species of the Caribbean coral reef. The identity of the polyP granules was confirmed by energy-dispersive spectroscopy (EDS) and by the fluorescence properties of the granules. Microscopy images revealed that a large proportion of microbial cells associated with sponge hosts contained intracellular polyP granules. Cyanobacterial symbionts cultured from sponges were shown to accumulate polyP. We also amplified polyphosphate kinase (ppk) genes from sponge DNA and confirmed that the gene was expressed. Based on these findings, we propose here a potentially important phosphorus (P) sequestration pathway through symbiotic microorganisms of marine sponges. Considering the widespread sponge population and abundant microbial cells associated with them, this pathway is likely to have a significant impact on the P cycle in benthic ecosystems.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium phosphate glass, Type 45
Sigma-Aldrich
DAPI, for nucleic acid staining