Merck
Clodronate inhibits tumor angiogenesis in mouse models of ovarian cancer.
Cancer biology & therapy (2014-05-21)
Nicole M Reusser, Heather J Dalton, Sunila Pradeep, Vianey Gonzalez-Villasana, Nicholas B Jennings, Hernan G Vasquez, Yunfei Wen, Rajesh Rupaimoole, Archana S Nagaraja, Kshipra Gharpure, Takahito Miyake, Jie Huang, Wei Hu, Gabriel Lopez-Berestein, Anil K Sood
ABSTRAKT
Bisphosphonates have been shown to inhibit and deplete macrophages. The effects of bisphosphonates on other cell types in the tumor microenvironment have been insufficiently studied. Here, we sought to determine the effects of bisphosphonates on ovarian cancer angiogenesis and growth via their effect on the microenvironment, including macrophage, endothelial and tumor cell populations. Using in vitro and in vivo models, we examined the effects of clodronate on angiogenesis and macrophage density, and the overall effect of clodronate on tumor size and metastasis. Clodronate inhibited the secretion of pro-angiogenic cytokines by endothelial cells and macrophages, and decreased endothelial migration and capillary tube formation. In treated mice, clodronate significantly decreased tumor size, number of tumor nodules, number of tumor-associated macrophages and tumor capillary density. Clodronate is a potent inhibitor of tumor angiogenesis. These results highlight clodronate as a potential therapeutic for cancer.