Merck
G-quadruplexes within prion mRNA: the missing link in prion disease?
Nucleic acids research (2014-07-18)
René C L Olsthoorn
ABSTRAKT
Cellular ribonucleic acid (RNA) plays a crucial role in the initial conversion of cellular prion protein PrP(C) to infectious PrP(Sc) or scrapie. The nature of this RNA remains elusive. Previously, RNA aptamers against PrP(C) have been isolated and found to form G-quadruplexes (G4s). PrP(C) binding to G4 RNAs destabilizes its structure and is thought to trigger its conversion to PrP(Sc). Here it is shown that PrP messenger RNA (mRNA) itself contains several G4 motifs, located in the octarepeat region. Investigation of the RNA structure in one of these repeats by circular dichroism, nuclear magnetic resonance and ultraviolet melting studies shows evidence of G4 formation. In vitro translation of full-length PrP mRNA, naturally harboring five consecutive G4 motifs, was specifically affected by G4-binding ligands, lending support to G4 formation in PrP mRNA. A possible role of PrP binding to its own mRNA and the role of anti-prion drugs, many of which are G4-binding ligands, in prion disease are discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %