Merck
  • Antiviral therapy improves post-hepatectomy survival in patients with hepatitis B virus-related hepatocellular carcinoma: a prospective-retrospective study.
Antiviral therapy improves post-hepatectomy survival in patients with hepatitis B virus-related hepatocellular carcinoma: a prospective-retrospective study.
Alimentary pharmacology & therapeutics (2014-11-22)
C C N Chong, G L H Wong, V W S Wong, P C T Ip, Y S Cheung, J Wong, K F Lee, P B S Lai, H L Y Chan
ABSTRAKT
The effect of antiviral therapy on the post-hepatectomy long-term survival in patients with hepatitis B virus (HBV)-related hepatocellular carcinoma (HCC) remains uncertain. To evaluate the effect of antiviral therapy on post-hepatectomy survival and recurrence in patients with HBV-related HCC. This was a prospective-retrospective study of a total of 404 patients who underwent hepatectomy for HBV-related HCC in a tertiary academic hospital. Data on patient and tumour characteristics, tumour recurrence, treatment for recurrence and survival were compared between antiviral and no antiviral groups. Patient's and tumour characteristics were comparable between the two groups, except a higher proportion of patients with cirrhosis in the antiviral group. With a mean follow-up time of 52.4 months, antiviral group had a better 5-year overall survival (66.7% vs. 56.0%, P = 0.001) while there was no significant difference in the 5-year disease-free survival (44.7% vs. 38.1%, P = 0.166). Use of antiviral therapy was associated with better liver function reserve at the time of recurrence and a greater proportion of patients could receive curative treatment for recurrence (38.5% vs. 24.3%, P = 0.041). There was no significant different in the hazard ratios of patients who started antiviral therapy before or after operation (P = 0.054). Use of antiviral therapy improves the long-term post-hepatectomy survival in patients with HBV-related HCC. With a better liver function reserve at the time of recurrence, a greater proportion of patients in antiviral group could receive curative treatment for recurrence.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Aspiryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard