Merck
Increased 68Ga-DOTATATE uptake in PET imaging discriminates meningioma and tumor-free tissue.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2015-01-31)
Walter Rachinger, Veit M Stoecklein, Nicole A Terpolilli, Alexander R Haug, Lorenz Ertl, Julia Pöschl, Ulrich Schüller, Christian Schichor, Niklas Thon, Jörg-Christian Tonn
ABSTRAKT
Meningiomas are known to express somatostatin receptor 2 (SSTR2). PET using the SSTR2 analog (68)Ga-DOTATATE has recently been introduced for imaging of meningiomas. However, a systematic correlation between (68)Ga-DOTATATE uptake, SSTR2 expression, and histology (including tumor-free scar tissue) is still lacking. For elucidation, we conducted this prospective study. Twenty-one adult patients with primary (n = 12) or recurrent (n = 9) meningiomas were prospectively enrolled. Preoperative MR imaging and (68)Ga-DOTATATE PET scans were fused and used for a spatially precise neuronavigated tissue-sampling procedure during tumor resection. Histopathologic diagnosis included immunohistochemical determination of SSTR2 expression. At each individual sampling site, the maximum standardized uptake value (SUVmax) of (68)Ga-DOTATATE was correlated with MR imaging findings, histology, and semiquantitative SSTR2 expression. One hundred fifteen samples (81 tumor, 34 tumor-free) were obtained. There was a significant positive correlation between SUVmax and SSTR2 expression. Receiver-operating characteristic analysis revealed a threshold of 2.3 for SUVmax to discriminate between tumor and nontumoral tissue. Regarding the detection of tumor tissue, PET imaging showed a higher sensitivity (90% vs. 79%; P = 0.049), with specificity and positive predictive values similar to MR imaging, for both de novo and recurrent tumors. (68)Ga-DOTATATE uptake correlates with SSTR2 expression and offers high diagnostic accuracy to delineate meningioma from tumor-free tissue even in recurrent tumors after previous therapy. Our findings substantiate an important role for (68)Ga-DOTATATE PET in meningioma management.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems