Merck
  • Effects of stress reduction on cardiovascular risk factors in type 2 diabetes patients with early kidney disease - results of a randomized controlled trial (HEIDIS).
Effects of stress reduction on cardiovascular risk factors in type 2 diabetes patients with early kidney disease - results of a randomized controlled trial (HEIDIS).
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association (2014-05-07)
S Kopf, D Oikonomou, M Hartmann, F Feier, V Faude-Lang, M Morcos, H-U Häring, W Herzog, A Bierhaus, P M Humpert, P P Nawroth
ABSTRAKT
Current guidelines for the treatment of type 2 diabetes focus on pharmacological treatment of glucose and cardio-vascular risk factors. The aim of this prospective randomized controlled intervention study was to examine the effects of a psychosocial intervention on clinical endpoints and risk factors in patients with type 2 diabetes and early diabetic kidney disease.110 patients were randomized to receive an 8-week mindfulness-based stress reduction (MBSR) training (n = 53) compared to standard care (n = 57). The study was carried out open-labelled and randomization was performed computer-generated in a 1:1 ratio. Primary outcome of the study was the change in urinary albumin excretion (albumin-creatinine-ratio, ACR); secondary outcomes were metabolic parameters, intima media thickness (IMT), psychosocial parameters and cardiovascular events.89 patients (42 in control group and 47 in intervention group) were analysed after 3 years of follow-up. After 1 year, the intervention group showed a reduction of ACR from 44 [16/80] to 39 [20/71] mg/g, while controls increased from 47 [16/120] to 59 [19/128] mg/g (p = 0.05). Parallel to the reduction of stress levels after 1 year, the intervention-group additionally showed reduced catecholamine levels (p < 0.05), improved 24 h-mean arterial (p < 0.05) and maximum systolic blood pressure (p < 0.01), as well as a reduction in IMT (p < 0.01). However, these effects were lost after 2 and 3 years of follow-up.This is the first study to show that a psychosocial intervention improves cardiovascular risk factors in high risk type 2 diabetes patients. Trial-Registration: NCT00263419 http://clinicaltrials.gov/ct2/show/NCT00263419 TRIAL REGISTRATION: clinicaltrials.gov-Identifier: NCT00263419.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material