Merck
  • Enhanced electrical responsiveness in the cerebral cortex with oral melatonin administration after a small hemorrhage near the internal capsule in rats.
Enhanced electrical responsiveness in the cerebral cortex with oral melatonin administration after a small hemorrhage near the internal capsule in rats.
Journal of neuroscience research (2014-06-26)
Yoshitomo Ueda, Tadashi Masuda, Akimasa Ishida, Sachiyo Misumi, Yuko Shimizu, Cha-Gyun Jung, Hideki Hida
ABSTRAKT
Intracerebral hemorrhage (ICH) can cause direct brain injury at the insult site and indirect damage in remote brain areas. Although a protective effect of melatonin (ML) has been reported for ICH, its detailed mechanisms and effects on remote brain injury remain unclear. To clarify the mechanism of indirect neuroprotection after ICH, we first investigated whether ML improved motor function after ICH and then examined the underlying mechanisms. The ICH model rat was made by collagenase injection into the left globus pallidus, adjacent to the internal capsule. ML oral administration (15 mg/kg) for 7 days after ICH resulted in significant recovery of motor function. Retrograde labeling of the corticospinal tract by Fluoro-Gold revealed a significant increase in numbers of positive neurons in the cerebral cortex. Immunohistological analysis showed that ML treatment induced no difference in OX41-positive activated microglia/macrophage at day 1 (D1) but a significant reduction in 8-hydroxydeoxyguanosin-positive cells at D7. Neutral red assay revealed that ML significantly prevented H2 O2 -induced cell death in cultured oligodendrocytes and astrocytes but not in neurons. Electrophysiological response in the cerebral cortex area where the number of Fluoro-Gold-positive cells was increased was significantly improved in ML-treated rats. These data suggest that ML improves motor abilities after ICH by protecting oligodendrocytes and astrocytes in the vicinity of the lesion in the corticospinal tract from oxidative stress and causes enhanced electrical responsiveness in the cerebral cortex remote to the ICH pathology.