Merck
Effect of chocolate and Propolfenol on rabbit spermatogenesis and sperm quality following bacterial lipopolysaccharide treatment.
Systems biology in reproductive medicine (2014-05-03)
Giulia Collodel, Elena Moretti, Maria Teresa Del Vecchio, Marco Biagi, Raffaella Cardinali, Lucia Mazzi, Gabriele Brecchia, Margherita Maranesi, Daniela Manca, Cesare Castellini
ABSTRAKT
The aims of the study were to evaluate the effects of chocolate and propolis-enriched diets on rabbit spermatogenesis, sperm motility, and ultrastructure following bacterial lipopolysaccharide (LPS) treatment. Thirty-two New Zealand White rabbits were divided into four groups. The LPS-Propolfenol(®) group received propolis (500 mg/kg/day) in their diet for 15 days, while the LPS-chocolate group was fed 70% cacao chocolate (1 g/1 kg/day) for the same period. Following the diet treatments, rabbits in the LPS-Propolfenol(®) and LPS-chocolate groups, and an LPS group received a single intraperitoneal dose of 50 μg/kg LPS, and the control group received only saline. Kinematic sperm traits were evaluated with a computer assisted sperm analyzer (CASA) system, and ultrastructural characteristics were examined by transmission electron microscopy (TEM). Testicular and epididymal tissues were observed by light microscopy and TEM and multiplex real time reverse transcriptase-polymerase chain reaction (RT-PCR) assay was used to detect and quantify toll-like receptor-4 (TLR-4) gene expression. The values of the analyzed semen parameters of rabbits treated with LPS-Propolfenol(®) and LPS-chocolate did not show any variations compared with the control group, but they were lower in rabbits treated only with LPS. Alterations observed in the testicular tissue of LPS treated-rabbits were not detected in specimens from the LPS-chocolate and LPS-Propolfenol(®) groups, which showed normal spermatogenesis. The TLR-4 mRNA expression was similar in controls, in LPS treated, and in LPS-chocolate groups, but it was significantly (p < 0.01) decreased in LPS-Propolfenol(®) rabbits. In conclusion, a chocolate and propolis-enriched diet showed a protective effect on the spermatogenetic process of buck rabbits following LPS treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Standardowa temperatura topnienia waniliny, United States Pharmacopeia (USP) Reference Standard
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Supelco
Vanillin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Epikatechina, primary reference standard