Merck
Association of hearing impairment and mortality in older adults.
The journals of gerontology. Series A, Biological sciences and medical sciences (2014-07-16)
Dane J Genther, Joshua Betz, Sheila Pratt, Steven B Kritchevsky, Kathryn R Martin, Tamara B Harris, Elizabeth Helzner, Suzanne Satterfield, Qian-Li Xue, Kristine Yaffe, Eleanor M Simonsick, Frank R Lin
ABSTRAKT
Hearing impairment (HI) is highly prevalent in older adults and is associated with social isolation, depression, and risk of dementia. Whether HI is associated with broader downstream outcomes is unclear. We undertook this study to determine whether audiometric HI is associated with mortality in older adults. Prospective observational data from 1,958 adults ≥70 years of age from the Health, Aging, and Body Composition Study were analyzed using Cox proportional hazards regression. Participants were followed for 8 years after audiometric examination. Mortality was adjudicated by obtaining death certificates. Hearing was defined as the pure-tone average of hearing thresholds in decibels re: hearing level (dB HL) at frequencies from 0.5 to 4kHz. HI was defined as pure-tone average >25 dB HL in the better ear. Of the 1,146 participants with HI, 492 (42.9%) died compared with 255 (31.4%) of the 812 with normal hearing (odds ratio = 1.64, 95% CI: 1.36-1.98). After adjustment for demographics and cardiovascular risk factors, HI was associated with a 20% increased mortality risk compared with normal hearing (hazard ratio = 1.20, 95% CI: 1.03-1.41). Confirmatory analyses treating HI as a continuous predictor yielded similar results, demonstrating a nonlinear increase in mortality risk with increasing HI (hazard ratio = 1.14, 95% CI: 1.00-1.29 per 10 dB of threshold elevation up to 35 dB HL). HI in older adults is associated with increased mortality, independent of demographics and cardiovascular risk factors. Further research is necessary to understand the basis of this association and whether these pathways might be amenable to hearing rehabilitation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydriodic acid, distilled, 57 wt. %, 99.999% trace metals basis
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%