Merck
Adenosine triphosphate acts as a paracrine signaling molecule to reduce the motility of T cells.
The EMBO journal (2014-05-21)
Chiuhui Mary Wang, Cristina Ploia, Fabio Anselmi, Adelaida Sarukhan, Antonella Viola
ABSTRAKT
Organization of immune responses requires exchange of information between cells. This is achieved through either direct cell-cell contacts and establishment of temporary synapses or the release of soluble factors, such as cytokines and chemokines. Here we show a novel form of cell-to-cell communication based on adenosine triphosphate (ATP). ATP released by stimulated T cells induces P2X4/P2X7-mediated calcium waves in the neighboring lymphocytes. Our data obtained in lymph node slices suggest that, during T-cell priming, ATP acts as a paracrine messenger to reduce the motility of lymphocytes and that this may be relevant to allow optimal tissue scanning by T cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture