Merck
  • Inter-individual variation in the effect of antiepileptic drugs in the intrahippocampal kainate model of mesial temporal lobe epilepsy in mice.
Inter-individual variation in the effect of antiepileptic drugs in the intrahippocampal kainate model of mesial temporal lobe epilepsy in mice.
Neuropharmacology (2014-12-03)
Sabine Klein, Marion Bankstahl, Wolfgang Löscher
ABSTRAKT
Despite more than 20 clinically approved antiepileptic drugs (AEDs), there remains a substantial unmet clinical need for patients with refractory (AED-resistant) epilepsy. Animal models of refractory epilepsy are needed for at least two goals; (1) better understanding of the mechanisms underlying resistance to AEDs, and (2) development of more efficacious AEDs for patients with refractory seizures. It is only incompletely understood why two patients with seemingly identical types of epilepsy and seizures may respond differently to the same AED. Prompted by this well-known clinical phenomenon, we previously evaluated whether epileptic rats respond differently to AEDs and discovered AED responsive and resistant animals in the same models. In the present study, we used the same approach for the widely used intrahippocampal kainate mouse model of mesial temporal lobe epilepsy. In a first step, we examined anti-seizure effects of 6 AEDs on spontaneous recurrent focal electrographic seizures and secondarily generalized convulsive seizures in epileptic mice, showing that the focal nonconvulsive seizures were resistant to carbamazepine and phenytoin, whereas valproate and levetiracetam exerted moderate and phenobarbital and diazepam marked anti-seizure effects. All AEDs seemed to suppress generalized convulsive seizures. Next we investigated the inter-individual variation in the anti-seizure effects of these AEDs and, in case of focal seizures, found responders and nonresponders to all AEDs except carbamazepine. Most nonresponders were resistant to more than one AED. Our data further validate the intrahippocampal kainate mouse model as a model of difficult-to-treat focal seizures that can be used to investigate the determinants of AED efficacy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Supelco
Carbamazepine, analytical standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
USP
Levetiracetam, United States Pharmacopeia (USP) Reference Standard
Levetiracetam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5,5-Diphenylhydantoin, ≥98%
Sigma-Aldrich
Levetiracetam, ≥98% (HPLC)
Supelco
Levetiracetam, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Fenytoina, United States Pharmacopeia (USP) Reference Standard
Phenytoin for system suitability, European Pharmacopoeia (EP) Reference Standard
Phenytoin, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenytoin, Pharmaceutical Secondary Standard; Certified Reference Material