Merck
  • Characterization of the potent odorants contributing to the characteristic aroma of Chinese green tea infusions by aroma extract dilution analysis.
Characterization of the potent odorants contributing to the characteristic aroma of Chinese green tea infusions by aroma extract dilution analysis.
Journal of agricultural and food chemistry (2014-08-05)
Ryoko Baba, Kenji Kumazawa
ABSTRAKT
The volatile fractions of three famous Chinese green tea cultivar infusions (Longjing, Maofeng, and Biluochun) were prepared by a combination of the adsorptive column method and the SAFE techniques. The aroma extract dilution analysis (AEDA) applied to the volatile fractions revealed 58 odor-active peaks with flavor dilution (FD) factors between 4(1) and 4(7). Forty-six of the odorants, which included six odorants that have not been reported in the literature in Chinese green tea (2-isopropyl-3-methoxypyrazine, 2-ethenyl-3,5-dimethylpyrazine, cis-4,5-epoxy-(E)-2-decenal, 4-ethylguaiacol, (E)-isoeugenol, and 3-phenylpropionic acid), were identified or tentatively identified by GC-MS and GC-O. Among the perceived odorants, 4-hydroxy-2,5-dimethyl-3(2H)-furanone, 3-hydroxy-4,5-dimethyl-2(5H)-furanone, coumarin, vanillin, geraniol, (E)-isoeugenol, and 2-methoxyphenol showed high FD factors in all of the cultivars, irrespective of the cultivar or harvesting season, suggesting that these seven odorants are essential for the aroma of Chinese green tea. On the other hand, the contents of the odorants, FD factors of which were uneven between the cultivars, were suggested to influence the characteristic aroma of each cultivar. In addition, the formation mechanism of (E)-isoeugenol, one of the odorants which have not been reported in the literature with a high FD factor common to all the cultivars, was investigated, and it was suggested that the (E)-isoeugenol content of the tea products has a close correlation with the manufacturing process of the tea leaves.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Coniferyl alcohol, 98%
Sigma-Aldrich
Cinnamyl alcohol, 98%
Sigma-Aldrich
Cinnamyl alcohol, ≥98%, FG