Merck
Inhibition of long non-coding RNA NEAT1 impairs myeloid differentiation in acute promyelocytic leukemia cells.
BMC cancer (2014-09-24)
Chengwu Zeng, Yan Xu, Ling Xu, Xibao Yu, Jingjing Cheng, Lijian Yang, Shaohua Chen, Yangqiu Li
ABSTRAKT
Acute promyelocytic leukemia (APL) is characterized by the reciprocal translocation t(15;17), which fuses PML with retinoic acid receptor alpha (RARα). Although PML-RARα is crucially important for pathogenesis and responsiveness to treatment, the molecular and cellular mechanisms by which PML-RARα exerts its oncogenic potential have not been fully elucidated. Recent reports have suggested that long non-coding RNAs (lncRNAs) contribute to the precise control of gene expression and are involved in human diseases. Little is known about the role of lncRNA in APL. We analyzed NEAT1 expression in APL samples and cell lines by real-time quantitative reverse transcription-PCR (qRT-PCR). The expression of PML-RARα was measured by Western blot. Cell differentiation was assessed by measuring the surface CD11b antigen expression by flow cytometry analysis. We found that nuclear enriched abundant transcript 1 (NEAT1), a lncRNA essential for the formation of nuclear body paraspeckles, is significantly repressed in de novo APL samples compared with those of healthy donors. We further provide evidence that NEAT1 expression was repressed by PML-RARα. Furthermore, significant NEAT1 upregulation was observed during all-trans retinoic acid (ATRA)-induced NB4 cell differentiation. Finally, we demonstrate the importance of NEAT1 in myeloid differentiation. We show that reduction of NEAT1 by small interfering RNA (siRNA) blocks ATRA-induced differentiation. Our results indicate that reduced expression of the nuclear long noncoding RNA NEAT1 may play a role in the myeloid differentiation of APL cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
Tretinoin, European Pharmacopoeia (EP) Reference Standard
USP
Tretynoina, United States Pharmacopeia (USP) Reference Standard