Merck
Controlled synthesis of Fe3O4/ZIF-8 nanoparticles for magnetically separable nanocatalysts.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-03-15)
Fei Pang, Mingyuan He, Jianping Ge
ABSTRAKT
Fe3O4/ZIF-8 nanoparticles were synthesized through a room-temperature reaction between 2-methylimidazolate and zinc nitrate in the presence of Fe3O4 nanocrystals. The particle size, surface charge, and magnetic loading can be conveniently controlled by the dosage of Zn(NO3)2 and Fe3O4 nanocrystals. The as-prepared particles show both good thermal stability (stable to 550 °C) and large surface area (1174 m(2) g(-1)). The nanoparticles also have a superparamagnetic response, so that they can strongly respond to an external field during magnetic separation and disperse back into the solution after withdrawal of the magnetic field. For the Knoevenagel reaction, which is catalyzed by alkaline active sites on external surface of catalyst, small Fe3O4/ZIF-8 nanoparticles show a higher catalytic activity. At the same time, the nanocatalysts can be continuously used in multiple catalytic reactions through magnetic separation, activation, and redispersion with little loss of activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Diethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Diethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Supelco
Diethylene glycol, analytical standard
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Cyanoacetamide, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Phenylacetic acid, 99%